missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Raji Whole Cell Lysate

Catalog No. NB800PC11
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB800PC11 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB800PC11 Supplier Novus Biologicals™ Supplier No. NB800PC11

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Raji Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.

Specifications

For Use With (Application) SDS-Page, Western Blot
Tissue Blood Lymph
Quantity 0.5 mg
Storage Buffer 1X SDS-PAGE Sample Buffer [0.05mM Tris HCl, 2% SDS, 10% Glycerol, 0.1% bromophenol blue, pH 6.8]
Test Specificity Raji whole cell extract lysate
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.

For Research Use Only