missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Neuro2a Whole Cell Lysate

Catalog No. NBP219017
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219017 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219017 Supplier Novus Biologicals™ Supplier No. NBP219017

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Neuro2a Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Immunoprecipitation.

Specifications

For Use With (Application) Immunoprecipitation
Tissue Brain
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), KCl, Sucrose, Sodium deoxycholate, and NP-40 (without denaturing agents).
Purity Protein
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.

For Research Use Only