missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Jurkat Cell Lysate

Catalog No. NBP225046
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225046 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225046 Supplier Novus Biologicals™ Supplier No. NBP225046
The Jurkat Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.

Specifications

Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Host Species Human
Content And Storage -80°C in the dark. Avoid freeze-thaw cycles.

For Research Use Only