Learn More
Novus Biologicals™ Daudi Cell Lysate
Catalog No. NBP225047
Change view
Click to view available options
Quantity:
0.1 mg
Catalog No. Quantity
NBP225047 0.1 mg
1 options
Catalog No. NBP225047 Supplier Novus Biologicals™ Supplier No. NBP225047
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Daudi Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
Specifications
Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Content And Storage -80°C in the dark. Avoid freeze-thaw cycles.
For Research Use Only