missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ L-Buffer Chamber for XCell SureLock™ and XCell II™ Mini-Cells DFS Item

XCell SureLock and XCell II Mini-Cells

Supplier:  Invitrogen™ EI0013

Numéro de catalogue. EI0013


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Spécifications

Spécifications

Store at room temperature.
Mini
Lower Buffer Chamber
Room Temperature
XCell SureLock™ Mini
XCell II, XCell SureLock
1 Each
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.