missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XCell II™ Blot Module

Catalog No. EI9051
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EI9051 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EI9051 Supplier Invitrogen™ Supplier No. EI9051
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Anode core, cathode core, sponge pads (8/pack), and instruction manual.

Easy transfer of proteins or nucleic acids from mini-gels to membranes

Fits neatly into the XCell SureLock™ and XCell II™ Mini-Cells in place of the gel/buffer core assembly. It requires less than 200mL of transfer buffer for Western, Southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 x 9cm.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Product Line XCell II
For Use With (Equipment) XCell SureLock™ Mini and Mini Gel Tank
Gel Compatibility Bolt Bis-Tris Plus Gel, NuPAGE Gel, Novex Mini Gel
Capacity Up to 2 mini-gels
Content And Storage The XCell II™ Blot Module includes anode core, cathode core, sponge pads (8/pack), and instruction manual.
Running Dimension Vertical
Product Type Blot Module
Mode of Transfer Wet
Shipping Condition Room Temperature
Gel Size Mini (8 cm x 8 cm)
Quantity 1 unit
Show More Show Less
Are Bolt gels compatible with transfer devices from other suppliers?

Yes. While we would prefer that you use our devices, Bolt gels can also be transferred using devices from Bio-Rad, including the Mini Trans-Blot Cell, Trans-Blot SD Semi-Dry Transfer Cell, or Trans-Blot Turbo Transfer System.

How can I improve transfer efficiency for larger proteins during western blotting?

Here are some options for obtaining more efficient transfer for larger proteins:

1) Pre-equilibrate the gel with 0.02 to 0.04% SDS in 2X transfer buffer without methanol for 10 min before assembling the sandwich.

2) Increase the blotting time incrementally (in 15 min intervals).

3) Add 0.01% or 0.02% SDS to the transfer buffer to help facilitate the migration of the protein out of the gel.

4) Decrease the methanol content in the transfer buffer.

5) Switch to a more appropriate lower-percentage gel. A lower-percentage gel may allow better transfer than a higher-percentage gel.

Do I have to run the XCell Blot Module under cold conditions?

No. The solution placed in the outer chamber serves to dissipate the heat generated during blotting. Water is usually used for this purpose. The recommended transfer conditions generate only a minor heat increase, so it is not necessary to run the unit in an ice bucket or to place it in a cold room. However if you are working with very heat-sensitive proteins, you may wish to do so.

How can I remove residual build-up in the XCell II Blot Module?

Build-up can be removed with 50% nitric acid. Make a solution of 50% nitric acid in deionized water and carefully apply it to areas inside the blot module until residual build-up is removed. Do not submerge the blot module or soak overnight. Use gloves when preparing the solution. Afterwards, rinse the module thoroughly at least three times in fresh deionized water. This treatment should not harm the plastic.

How can residual build-up in the XCell II Blot Module be removed?

Build-up can be removed with 50% nitric acid. Make a solution of 50% nitric acid in deionized water and carefully apply it to areas inside the blot module until residual build-up is removed. Do not submerge the blot module or soak overnight. Use gloves when preparing the solution. Afterwards, rinse the module thoroughly at least three times in fresh deionized water. This treatment should not harm the plastic.

Can Invitrogen gels be blotted with a Invitrogen Semi-Dry Blotter?

The NuPAGE Invitrogen Bis-Tris Gels do not transfer efficiently using a Invitrogen Semi-Dry Blotter as compared to blotting with XCell II Blot Module.
If you decide to use Invitrogen Semi-Dry Blotter for NuPAGE Invitrogen Bis-Tris Gels, use the protocol provided below to ensure efficient transfer of proteins.

1) Prepare 100 mL of 2X NuPAGE Transfer Buffer from 20X NuPAGE Transfer Buffer as follows:
NuPAGE Transfer Buffer (20X) 10.0 mL
NuPAGE Antioxidant (for reduced sample) 0.1 mL
Methanol 10.0 mL
Deionized water 79.9 mL
Total Volume 100 mL
If you are blotting large proteins, please see the Note below.

2) Soak the filter paper and transfer membrane in the transfer buffer.
If you are using Invitrogen pre-cut membrane/filter sandwiches, use three filter papers (0.4 mm/filter in thickness) on each side of the gel or membrane.
If you are not using the Invitrogen pre-cut membrane/filter sandwiches, use two thick filter papers.

3) Assemble the gel/membrane/filter paper sandwich on top of the anode plate as follows:
filter paper
filter paper
filter paper
membrane
gel
filter paper
filter paper
filter paper

4) Perform the transfer at 15 V (constant) for 15 min if you are using the Bio-Rad Trans-Blot Semi-Dry Transfer Cell. For any other semi-dry transfer cell, follow the manufacturer's recommendations.

Note: For transfer of large proteins (>100 kDa), pre-equilibrate the gel in 2X NuPAGE Transfer Buffer (without methanol) containing 0.02-0.04% SDS for 10 min before assembling the sandwich.

How do I destain proteins on a PVDF membrane that were stained with SimplyBlue SafeStain?

After staining with SimplyBlue SafeStain, use deionized water for the less strongly retained protein bands on the PVDF membrane.

Increasing methanol or ethanol concentrations up to 70% should destain any remaining bands. You can leave the membrane in the destain indefinitely.

During Western transfer conditions using constant voltage, what would cause the current reading to drop much lower than the expected starting current?

There are three common explanations:

1) The buffer was accidentally made too dilute, which increases resistance and lowers conductivity and current. Check the transfer buffer and its reagent components, remake, or redilute.

2) The circuit is broken or impeded, as in the case of a corroded or broken electrode or malfunctioning power supply. Check the equipment.

3) There is a leak in the blot module. This is indicated by a drastic decrease in current and in buffer volume within the module.

What causes the appearance of diffuse bands and swirling patterns on membranes after Western transfer?

The swirling and diffuse banding patterns are typical of molecules moving laterally before binding to the membrane during transfer. Three possible explanations are:

1) Poor contact between the gel and the membrane: the gel should contact the membrane across the entire surface for good capillary action. To ensure good contact and remove any bubbles, roll a glass pipette over the surface of each layer of the gel/membrane sandwich. Pipet some extra transfer buffer on the surface of each layer as the sandwich is being made. Also, the blotting pads need to be fully saturated with transfer buffer (push down with a gloved hand when they are submerged to ensure that there are no air bubbles).

2) Undercompression of the gel: The gel/membrane assembly should be held securely between the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with new ones.

3) Overcompression of the gel: A good indication of overcompression is if the gel has been excessively flattened. In the event that the sandwich is overcompressed, remove enough pads so that the blot module can be closed without exerting excess pressure on the gel and membrane. NOTE: The height of the uncompressed pads should be 0.5-1.0 cm above the level of the sealing gasket.

What is the 54 kDa band observed in human tissue lysates when I use WesternBreeze kits?

A number of human alkaline phophatases that could be consuming the WesternBreeze substrate have a molecular weight close to 50 kDa. These would be detected as a band of approximately 54 kDa. These proteins are known to dimerize, and can show as a band of approximately 110 kDa in native gels.

Do you have any tips to improve results when Western transferring two gels at the same time?

Less-than-optimal transfer in the second gel is not uncommon and frequently requires adjustments in protocol. To achieve similar transfer efficiencies in the two gels you can transfer them one at a time or consider the following suggestions:

Make sure that the blot module is not overfilled with buffer, i.e., fill to point where pads are just covered and no higher. If the buffer level is too high, some current will bypass the gels. If it is overfilled, shorting, arcing, and other problems can occur.

Use antioxidant when transferring reduced proteins.

Increase transfer time by 30 to 60 minutes. The longer run time allows the slower moving proteins time to move out of the gel.

As proteins are being driven out of the gel by the SDS on their surface, it is important that enough is retained to keep them mobile. However, the methanol that is included in the transfer buffer will remove some of the SDS, and the higher the methanol concentration, the more SDS is washed off of the protein. Methanol is included in order to increase binding to nitrocellulose membranes via hydrophobic interactions. However, if using PVDF or nylon membranes, less or even no methanol may be required. 20% methanol is a good starting point for most situations In the event that the transfer is less than ideal, methanol levels should be reduced by at least 50%. The optimal percentage should be determined empirically. Something to keep in mind is that with increased mobility, the proteins may just move straight though the first membrane. In this case, a second membrane may trap these samples.

It is possible to BRIEFLY (1 min) soak the second gel in transfer buffer with 0.01 to 0.02% SDS before assembling the sandwich. This must be a short soak or the proteins will tend to diffuse out of the gel. Adding 0.01-0.02% SDS to the transfer buffer and using it for the transfer will also aid in protein mobility.

Halfway through the transfer process (30 min), swap the front gel and back gel in the blotter. This is fairly easy to do if you just swap pad-paper-gel-membrane-pad sections.

Another recommendation: if you must transfer simultaneously, and want to be able to compare results of both blots, use an internal control in both gels to get an idea of the relative transfer efficiency and to afford some normalization of the results.

I am transferring a Tris-Glycine gel using constant voltage and the current reading is way over the expected starting current. Can you offer some suggestions?

The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.

I ran my protein under native conditions on a Tris-Glycine gel. It has a pI that is higher than the pH of the Tris-Glycine transfer buffer. Can you offer some tips for transferring it?

- Increase the pH of Tris-Glycine transfer buffer to 9.2, allowing all the proteins below pI 9.2 to transfer towards the anode electrode.
- Use the Tris-Glycine transfer buffer and place a membrane on both sides of the gel. If there are any proteins that are more basic than the pH of the transfer buffer, they will be captured on the extra membrane placed on the cathode side of the gel. Both membranes can then be developed in the same manner.
- Prior to blotting, incubate the gel for 15 minutes in Tris-Glycine transfer buffer containing 0.1% SDS. The small amount of SDS will give the proteins enough charge to move unidirectionally towards the anode and in most cases, should not denature the protein. Proceed with the transfer using regular Tris-Glycine transfer buffer.

I had problems transferring my larger-molecular weight proteins from my NuPAGE gel. Can you please offer some suggestions?

For proteins larger than 100 kDa, we recommend pre-equilibrating the gel in 2X NuPAGE Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X NuPAGE transfer buffer containing methanol and 0.01% SDS.

What causes empty spots on my membrane after transfer?

Here are possible causes and solutions:

- Presence of air bubbles between the gel and the membrane preventing the transfer of proteins. Be sure to remove all air bubbles between the gel and membrane by rolling a glass pipette over the membrane surface.
- Expired or creased membranes used. Use fresh, undamaged membranes.

I performed a western transfer and see the appearance of diffuse bands and swirling patterns on the membrane. What could have happened?

The swirling and diffuse banding patterns are typical of molecules moving laterally before binding to the membrane during transfer. Here are possible causes and solutions:

- Poor contact between the gel and the membrane: The gel should be attached to the membrane through capillary action. To ensure that this happens, make sure that you roll over the surface of each layer of the gel/membrane sandwich with a glass pipette to ensure good contact between the gel and the membrane. It is helpful to use a disposable pipette to place some extra transfer buffer on the surface of each layer as the sandwich is being made. Also, the pads need to be fully saturated (push down with gloved hand when they are placed in transfer buffer to make sure there are no air bubbles.)
- Under-compression of the gel: The gel/membrane assembly should be held securely between the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones.
- Over-compression of the gel: A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and membrane.
Note: The height of the uncompressed pads should be 0.5-1.0 cm above the level of the sealing gasket.

When I perform a western transfer, the power supply shuts off in the middle of the transfer. What is wrong?

Here are possible causes and solutions:

- High ionic strength of the transfer buffer. Prepare the buffer as described in the manual.
- Power supply is operating at a current close to the current limit of the power supply. Use a power supply with higher limits.

After a western transfer, I noticed that a significant amount of protein remained in the gel indicated by staining of the gel after transfer. What should I do?

Here are possible causes and solutions:

- Too short a transfer time: Increase the blotting time by 15 minute increments.
- Inappropriate gel type: Check the percentage of the gel used and switch to a higher percentage gel.
- Inappropriate amount of SDS: Add 0.01-0.02% SDS to the transfer buffer to facilitate migration of the protein out of the gel.
- Inappropriate methanol content: Decrease the amount of methanol in the transfer buffer.
Note: Higher molecular weight proteins usually do not transfer completely as compared to mid to low molecular weight proteins.

After a western transfer, I noticed that a significant amount of protein had passed through the membrane indicated by the presence of proteins on the second membrane. Can you please help?

Here are possible causes and solutions:

- Too long a transfer tim: Shorten the transfer time by 15 minute increments.
- Inappropriate amount of SDS: Do not include any SDS in the transfer buffer.
- Inappropriate methanol content: Add additional methanol to the transfer buffer to increase the binding capacity of the membrane.
- Inappropriate gel type: Check the percentage of the gel used and switch to a higher percentage gel.
- Sample overloaded: Decrease the sample load.
- Finally, if using nitrocellulose membrane, switch to PVDF which has a higher binding capacity.

I performed a western transfer and none of my proteins transferred to the membrane. Can you offer some tips?

It is possible that the gel/membrane sandwich was assembled in the reverse direction such that the proteins have migrated out into the buffer. Assemble the blot sandwich in the correct order using instructions provided in the manual.

During western transfer conditions using constant voltage, what would cause the actual current to greatly exceed the expected starting current?

The most common cause of abnormally high current is the buffer. If the buffer is too concentrated, this leads to increased conductivity and higher current. High current may also occur if Tris-HCl was accidentally substituted for the Tris base required in the transfer buffer. Tris-HCl results in a low buffer pH and leads to increased conductivity and current, and, subsequently, overheating. Check the transfer buffer and its reagent components, re-dilute, or remake the buffer.

I am transferring my gel using constant voltage and the current reading has dropped much lower than the expected starting current. What could have happened?

Here are possible causes and solutions:

- The buffer was accidentally made too dilute, therefore increasing resistance and thus lowering conductivity and current: Check the transfer buffer and its reagent components and then re-dilute it or remake it.
- The circuit is broken or impeded, as in the case of a corroded or broken electrode or malfunctioning power supply: Check the equipment.
- There is a leak in the blot module (this is indicated by a drastic decrease in current and in buffer volume within the module): Ensure that the inner buffer chamber is filled sufficiently so that the wells are covered with buffer.
- Tape at the bottom of the gel cassette was not removed: Double check that the tape on the bottom of the gel has been removed.

What conditions do you recommend for overnight blotting using the XCell II Blot Module?

For overnight blotting, perform transfer in the cold room with low power to prevent overheating. Transfer at constant voltage of 10-15 V overnight. Depending on the transfer efficiency, adjust the transfer conditions accordingly.

What are your recommendations for cleaning of the XCell II Blot Module?

We recommend rinsing the blot module with deionized water after use. To clean any residual build-up in the blot module, apply 50% nitric acid in deionized water to areas inside the blot module until residual build-up is removed. Once the build-up is removed, rinse the module at least three times in deionized water. Do not submerge the blot module or soak overnight in nitric acid. Please see page 17 of the XCell II Blot Module Manual.

Note: Use gloves when preparing the nitric acid solution.

What will happen if the XCell II Blot Module outer buffer chamber is accidentally filled with transfer buffer instead of water?

This is perfectly acceptable as the water in the outer buffer chamber only serves as a coolant. We recommend adding deionized water to the outer buffer chamber to avoid exposure of the mini-cell to methanol as the mini-cell is susceptible to methanol.

Do you offer replacement parts for the XCell II Blot Module?

Except for the sponge pads (Cat. No. EI9052), none of the other parts are available separately.

Should I clean the sponge pads after each use and how should I do it?

We recommend cleaning the sponge pads after each use by rinsing with deionized water and squeezing the water out of the sponge pad 3-5 times.

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.


For Research Use Only. Not for use in diagnostic procedures.