missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternBreeze™ Chemiluminescent Kit, anti-rabbit

Catalog No. WB7106
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WB7106 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WB7106 Supplier Invitrogen™ Supplier No. WB7106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots)

Detection is accomplished with a ready-to-use CDP-Star™ chemiluminescent substrate for alkaline phosphatase. Protein bands can be captured either by X-ray film or CDP-Star™ compatible imaging system.

WesternBreeze™ Chemiluminescent Kit offers:

  • High specificity, clean background
  • Ultra-sensitivity-femtogram levels detectable
  • Long-lasting signals-up to 5 days
  • Results in less than 3 hours
  • Permanent photographic image

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Product Line WesternBreeze
Detection Method Chemiluminescence
Membrane Compatibility Nitrocellulose, PVDF
Product Type Western Blot Kit
Content And Storage The WesternBreeze™ Chemiluminescent Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.
Shipping Condition Wet Ice
Substrate Type AP (Alkaline Phosphatase) Substrate
Reactivity Rabbit
Cross Reactivity Rabbit
Quantity 1 kit
Why is the actual band size on a western blot different from the predicted size of the protein?

Western blotting is based on the separation of proteins by their size on a gel. However, migration of proteins through the gel matrix is also affected by other factors, which may cause the observed band size to be different from the predicted size.

Common causes are:
-Post-translational modification; for example phosphorylation and glycosylation increase the size of the protein
-Post-translation cleavage; many proteins are synthesized as precursor proteins, and then cleaved to give the active form
-Multimers, for example dimerization of a protein. This is usually prevented under reducing conditions, although strong interactions can result in the appearance of higher bands
-Splice variants; alternative splicing may result in different sized proteins being produced from the same gene
-Relative charge; the composition of amino acids (charged vs. non-charged)

What are the standard lysis buffers used with mammalian cells for detection of protein expression by immunoprecipitation (IP) or Western blot analysis?

The most commonly used buffer is RIPA Buffer with SDS. We offer RIPA Buffer (Cat. Nos. 89900 and 89901). We also offer the Pierce IP Lysis buffer (Cat. Nos. 87787 and 87788) as well as M-PER (Cat. Nos. 78501, 78503, and 78505).

What conditions do you recommend for overnight Western transfer?

Doing an overnight Westerm transfer is not the preferred method but can be done. The power should be lowered and the buffer should be chilled or the unit should be placed in the cold room to prevent overheating. You may try an overnight transfer at 5-15 V and adjust accordingly. You may also wish to put a second membrane behind the first in order to bind any proteins that transfer through the first membrane. You can use both membranes for staining, immunoblotting, or analysis.

Can I purchase the Secondary Antibody solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?

The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007), but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.

Can I purchase the Blocker/Diluent and Antibody Wash solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?

Yes, you may purchase them as standalone products using the Cat. Nos. listed below:

- Cat. No. WB7001 (Blocker/Diluent A)
- Cat. No. WB7002 (Blocker/Diluent B)
- Cat. No. WB7050 (Combo pack containing Blocker/Diluents A & B)
- Cat. No. WB7003 (Antibody Wash)

Note: Cat. Nos. WB7001 and WB7002 may be ordered by clicking the Quick Order button located at the top right of any page on our website.

Can I purchase the Chemiluminescent Substrate and Chemiluminescent Substrate Enhancer from the WesternBreeze Chemiluminescent Detection Kits as standalone products?

Yes, you can purchase them as standalone products and below are the Cat. Nos.:

- Chemiluminescent Substrate, Cat. No. WP20002
- Chemiluminescent Substrate Enhancer, Cat. No. WP20003

How should I prepare my membranes for WesternBreeze Chemiluminescent detection?

For western blots, where proteins are freshly transferred from SDS-PAGE gels to nitrocellulose or PVDF membranes, washing the membranes twice for 5 mins with 20 mL of pure water is recommended to partially remove gel and transfer buffer components and weakly bound proteins. The membranes are then ready for the WesternBreeze Chemiluminescent Immunodetection protocol.

Alternatively, the washed membranes may be dried on a clean piece of filter paper in open air, by a stream of slightly warm air or under an infrared lamp. Properly dried membranes can be stored in a closed container at 4 degrees C for several days depending on the antigen loaded. Water-washed and dried nitrocellulose membranes are ready for the WesternBreeze Chemiluminescent Immunodetection protocol. However, water-washed and dried PVDF membranes require a re-wetting step in methanol, followed by two 20 mL water washes for 5 mins before proceeding to the WesternBreeze Chemiluminescent Immunodetection protocol.

For Native-PAGE western blot, a drying step, performed before any washing steps, is recommended to improve protein binding to the membrane. Once dried, nitrocellulose membranes should be washed twice with 20 mL water for 5 mins before proceeding to the WesternBreeze Chemiluminescent Immunodetection protocol. Dried PVDF membranes require a re-wetting step in methanol, followed by two 20 mL water washes for 5 minutes before proceeding to the WesternBreeze Chemiluminescent Immunodetection protocol.


For Research Use Only. Not for use in diagnostic procedures.