missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternBreeze™ Blocker/Diluent (Part A and B)

Catalog No. WB7050
Encompass
Change view
Click to view available options
Quantity:
80 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WB7050 80 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WB7050 Supplier Invitrogen™ Supplier No. WB7050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

80 mL of Blocker/Diluent (Part A); 80 mL of Blocker/Diluent (Part B)

Optimized, easy-to-use blocker and primary antibody diluent system

Yields low background/high signal western blot detection on nitrocellulose (NC) and polyvinylidene difluoride (PVDF) membranes. Sufficient reagents for 20 mini-blots.

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Product Line WesternBreeze
Membrane Compatibility Nitrocellulose, PVDF
Product Type Blocker and Diluent
Buffer Blotting Buffers
Content And Storage 80 ml of Blocker/Diluent (Part A);
80 ml of Blocker/Diluent (Part B).

Store at +4°C.
Quantity 80 mL
Why is the actual band size on a western blot different from the predicted size of the protein?

Western blotting is based on the separation of proteins by their size on a gel. However, migration of proteins through the gel matrix is also affected by other factors, which may cause the observed band size to be different from the predicted size.

Common causes are:
-Post-translational modification; for example phosphorylation and glycosylation increase the size of the protein
-Post-translation cleavage; many proteins are synthesized as precursor proteins, and then cleaved to give the active form
-Multimers, for example dimerization of a protein. This is usually prevented under reducing conditions, although strong interactions can result in the appearance of higher bands
-Splice variants; alternative splicing may result in different sized proteins being produced from the same gene
-Relative charge; the composition of amino acids (charged vs. non-charged)

What are the standard lysis buffers used with mammalian cells for detection of protein expression by immunoprecipitation (IP) or Western blot analysis?

The most commonly used buffer is RIPA Buffer with SDS. We offer RIPA Buffer (Cat. Nos. 89900 and 89901). We also offer the Pierce IP Lysis buffer (Cat. Nos. 87787 and 87788) as well as M-PER (Cat. Nos. 78501, 78503, and 78505).

Can I purchase the Blocker/Diluent and Antibody Wash solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?

Yes, you may purchase them as standalone products using the Cat. Nos. listed below:

- Cat. No. WB7001 (Blocker/Diluent A)
- Cat. No. WB7002 (Blocker/Diluent B)
- Cat. No. WB7050 (Combo pack containing Blocker/Diluents A & B)
- Cat. No. WB7003 (Antibody Wash)

Note: Cat. Nos. WB7001 and WB7002 may be ordered by clicking the Quick Order button located at the top right of any page on our website.

Can I purchase the Blocker/Diluent and Antibody Wash solutions from the WesternBreeze Chromogenic Detection kits as standalone products?

Yes, you may purchase them as standalone products using the Cat. Nos. listed below:

- Cat. No. WB7003 (Antibody Wash)
- Cat. No. WB7001 (Blocker/Diluent A)
- Cat. No. WB7002 (Blocker/Diluent B)
- Cat. No. WB7050 (Combo pack containing Blocker/Diluents A & B)

For Research Use Only. Not for use in diagnostic procedures.