Learn More
Invitrogen™ Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 DFS Item
Use with Novex Tricine Gels
Supplier:  Invitrogen™ LC1677
Catalog No. LC1677
Only null left
Includes: Novex Tricine SDS Running Buffer (10X, 500mL - LC1675) and Novex Tricine SDS Sample Buffer (2X, 20mL - LC1676)
Description
Description
In the tricine system developed by Schaegger and von Jagow (1), tricine replaces glycine in the running buffer. This results in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
Novex Tricine Gel
SDS PAGE Gel
Buffer Kit
The Novex™ Tricine SDS Buffer Kit includes Novex™ Tricine SDS Running Buffer (10X, 500 ml - LC1675) and Novex™ Tricine SDS Sample Buffer (2X, 20 ml - LC1676).
Novex
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.