Learn More
Invitrogen™ Porcine TNF alpha ELISA Kit
Description
Includes
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
The Porcine Tumor Necrosis Factor alpha (Por TNFα) ELISA quantitates Por TNFα in porcine serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Por TNFα1. Principle of the method The Pociner TNFαsolid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Accession Number | P23563 |
| Assay Range | 23.4-1500 pg/mL |
| Assay Sensitivity | <3 pg/mL |
| Content And Storage | Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Protein Family | Cytokines & Receptors |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Pig |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.