missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNF alpha Porcine Matched Antibody Pair

Catalog No. CSC1753
Encompass
Change view
Click to view available options
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CSC1753 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CSC1753 Supplier Invitrogen™ Supplier No. CSC1753

Includes

Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.

Requires

Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).

Matched Antibody Pair

The Porcine Tumor Necrosis Factor alpha Antibody Pair Kit comes with pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of pig TNFα. Buffer reagents needed to complete the ELISA reaction are sold separate as a Buffer Kit for Antibody Pairs (Catalog No. CNB0011). The Assay Buffer included in this Buffer Kit can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.

Order Info

Shipping Condition: Wet Ice

Specifications

Conjugate HRP
Gene Accession No. P23563
Gene Alias MGC124630, RATTNF, TNF-alpha, Tnfa
Gene Symbols Tnf
Quantity 10 x 96 Tests
Regulatory Status RUO
Gene ID (Entrez) 397086
Includes Capture (coating) Antibody, Detection Antibody, Standard, Streptavidin-HRP
Target Species Pig
Content And Storage 2°C to 8°C
Product Type ELISA Kit
For Use With (Equipment) Colorimetric Microplate Reader
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.