missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA TMB Stabilized Chromogen

Catalog No. SB01
Encompass
Change view
Click to view available options
Quantity:
1 L
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01 25 mL
SB02 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB01 Supplier Invitrogen™ Supplier No. SB01
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with all sandwich ELISAs which utilize Horseradish Peroxidase (HRP) as detecting enzyme

  • Formulation: Liquid
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

Specifications

Quantity 25 mL
Content And Storage Solution. Store at 2-8°C.
Detection Method Colorimetric
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.