missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMB Solution (BLOT)
Description
May be used with horseradish peroxidase (HRP)-based immunodetection procedures
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping condition: Wet ice
Specifications
Specifications
| Content And Storage | Store at 2-8°C |
| Detection Method | Colorimetric |
| Product Type | TMB Solution |
| Substrate | TMB |
| Substrate Properties | Chemical Substrate |
| Substrate Type | HRP (Horseradish Peroxidase) Substrate |
| Quantity | 500 mL |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.