missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE-Urea Gels, 15%

Catalog No. EC6885BOX
Encompass
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
EC6885BOX 10-well
EC68852BOX 12-well
EC68855BOX 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EC6885BOX Supplier Invitrogen™ Supplier No. EC6885BOX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels. Novex TBE-Urea Gels have a shelf life of between 4 and 8 weeks depending upon gel type.

Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands.

Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands. These 15% gels are optimized for the analysis and purification of products ranging from 10–40 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and northern blot analysis.

Novex TBE-Urea Gels can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis. The compact gels are available in 6%, 10%, or 15% formats and are designed to run on the XCell SureLock™ Mini-Cell.

Advantages of TBE-Urea Gels:

  • Contains 7M urea for maximum denaturation
  • Excellent resolution for fast size and purity confirmations of DNA or RNA oligos
  • High resolution of short single-strand oligonucleotides
  • Requires ∼10% sample concentration and volume of large or agarose gels
  • Accurate and reproducible results

Formulation

Novex TBE-Urea gels are made with high-purity reagents, including Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, APS, and 7M urea. Strict quality control ensures that gels and buffers are DNase- and RNase-free.

Recommended Buffers

For optimum performance, Novex™ TBE-Urea sample buffers and Novex™ TBE Running Buffer are strongly recommended for use with these gels. For analytical applications, Novex TBE-Urea Sample Buffer is recommended contains urea, the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage • Catalog number refers to a single box of 10 gels
• Novex™ TBE-Urea Gels have a shelf life of 4 to 8 weeks depending upon gel type

Store at 4°C.
Description Novex TBE-Urea Gels, 15%, 10-well
Gel Percentage 15%
Gel Type TBE-Urea
Separation Range 10 to 40 bp
Wells 10-well
Quantity 10 Gels/Box
Sample Type Single-stranded DNA Oligos/RNA
Shipping Condition Wet Ice
Product Type TBE Gel
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
I am getting high background and smeared bands on my TBE-urea gels. What should I do?

Here are some suggestions for your experiments:

- The RNA samples may have been degraded by RNases. Use standard precautions to prevent contamination.
- Make sure samples are heated for 3 minutes at 70°C just prior to loading. If this is not done, the oligonucleotides will not be fully denatured, which may result in a smeared background.
- Be sure to vigorously flush urea out of sample wells just prior to loading the sample. Urea will continually seep from the gel into the well. Urea is very dense and will force the sample into a ball.
- A sample volume over 10 µL may result in smearing. This volume is less than may be used with agarose gels.
- If ultrapure water (18 milliohms) is not used, smeared bands and high background may result.
- Check to make sure the running and sample buffers have been prepared and diluted correctly.
- Make sure to run the bromophenol blue until it reaches the slot. Stopping the run short can result in less than optimal results.

Can I use a sample buffer containing formamide with the Invitrogen TBE-urea system?

There are many sample buffer formulations used; however, we have found a distinct difference in the band appearance depending on the sample buffer composition. After evaluating urea, formamide, and various buffer systems, we found that the sharpest, flattest bands were obtained with a urea, Ficoll, and TBE buffer solution. Sample buffers made with formamide provided fuzzy, indistinct bands.

Can I stain my TBE gel or my TBE-urea gel? How?

Yes, for ethidium bromide staining, soak the gel in a 2 µg/mL solution of ethidium bromide in ultrapure water for 20 minutes. Destain by rinsing with three successive 10-minute rinses of ultrapure water. Visualize bands under UV light

I am planning to use a TBE polyacrylamide gel for electrophoresis of my nucleic acid fragment. How do I know if the size of my fragment is suitable for a particular gel you offer?

To help you choose the TBE polyacrylamide gel that will provide optimal separation results for your nucleic acid fragment, we have designed gel migration charts for each gel type offered. Please check the TBE gel migration charts on page 74 of the Novex Pre-Cast Gel Electrophoresis Guide (https://tools.thermofisher.com/content/sfs/manuals/electrophoresisguide_man.pdf).

How can I isolate DNA from Thermo Fisher Scientific TBE and TBE-Urea gels?

Here is the protocol you can follow:
(1) Run the gel. Locate the DNA of interest by autoradiography or by examination of ethidium bromide stained gels.
(2) Use a razor blade to cut out the upper 1/2 to 2/3 of the band of interest.
To recover a fragment of DNA identified by autoradiography, cut out from the X-ray film a small rectangle encompassing the image of the fragment. Align the film over the gel and cut out the segment of polyacrylamide bordered the rectangular hole in the film.
(3) Transfer the gel slice to a microcentrifuge tube. Use a sterile glass rod to crush the slice against the wall of the tube.
(4) Calculate the volume of the slice and add 1 to 2 volumes of elution buffer to the microcentrifuge tube.
Elution buffer: 0.5 M ammonium acetate, 10 mM magnesium sulfate, 1 mM EDTA (pH 8.0), 0.1% SDS
It is convenient if the volume of elution buffer is no greater than 0.5 mL since the eluted fragment of DNA can then be precipitated with ethanol in a single tube.
(5) Close the tube and incubate at 37 degrees C on a rotary platform. Small fragments of DNA (500 bases) are eluted in 3 to 4 hours.
(6) Centrifuge the sample at 12,000 g for 1 minute at 4 degrees C in a microcentrifuge. Transfer the supernatant to a fresh microcentrifuge tube, being careful to avoid transferring fragments of polyacrylamide.
(7) Add additional 0.5 mL volume of elution buffer to the pellet of polyacrylamide, vortex briefly, and re-centrifuge. Combine the two supernatants.
(8) Remove any remaining fragments of polyacrylamide by passing the supernatant through a disposable plastic column or a syringe barrel containing Whatman GF/C filter or packed siliconized glass wool.
(9) Add 2 volumes of ethanol at 4 degrees C and store the solution on the ice for 30 min. Recover the DNA by centrifugation at 12,000 g for 10 min at 4 degrees C in a microcentrifuge.
(10) Re-dissolve the DNA in 200 µL of Tris-EDTA (TE) buffer (pH 7.6), add 25 µL of 3M sodium acetate (pH 5.2) and re-precipitate the DNA with 2 volumes of ethanol as described in step 9. TE Buffer (pH 7.6): 10 mM Tris HCl (pH 7.6), 1mM EDTA (pH 8.0).
(11) Carefully rinse the pellet once with 70% ethanol and redissolve the DNA in TE buffer (pH 7.6) to a final volume of 10 µL.
(12) Check the amount and the quality of the fragment by polyacrylamide gel electrophoresis.

Can a sample buffer with formamide be used with the Invitrogen TBE-Urea system?

There are many sample buffer formulations used, however we have found a distinct difference in the band appearance depending on the sample buffer composition. After evaluating urea, formamide, and various buffer systems, we found that the sharpest, flattest bands were obtained with a urea, Ficoll, and TBE buffer solution. Sample buffers made with formamide resulted in fuzzy, indistinct bands.

What can be used to stain Invitrogen precast TBE-Urea or TBE gels?

(1) Ethidium bromide: Soak gel in a 2 µg/mL solution of ethidium bromide in ultrapure water for 20 min. Destain by rinsing with three successive 10-min rinses of ultrapure water. Visualize bands under UV light. The gel must be removed from the cassette prior to visualization of the DNA under a UV light. Because polyacrylamide quenches the fluorescence of ethidium bromide, it is not possible to detect bands that contain less than about 10 ng of DNA by this method. SilverXpress and SYBR stains will provide greater detection sensitivity.

(2) Invitrogen SilverXpress Stain: Follow the standard procedure from the instruction booklet for staining TBE or TBE-Urea gels.

(3) SYBR Green I/II Nucleic Acid Gel Stain: See the SYBR Green Staining Manual for protocol details.

What can be done to prevent abnormal band shapes (round bands instead of flat sharp bands) and poor resolution on a TBE-Urea Gel?

(1) Make sure urea is thoroughly flushed from sample wells just prior to loading samples. Urea will continuously diffuse into the wells and will force the sample into a ball, resulting in rounded bands rather than flat bands.

(2) Use the Invitrogen TBE-Urea sample buffer. We have found that our TBE-Urea sample buffer provides the sharpest, flattest bands on our gels.

(3) Load less sample onto the gel. For the flattest, sharpest bands and highest resolution, load between 0.16 and 0.33 µg (0.005 - 0.01 OD) per band. More than that will result in rounded bands.

What prevents high background and smeared bands on TBE-Urea gels?

(1) If running RNA samples, the samples may have been degraded by RNases. Use standard precautions to prevent contamination.

(2) Make sure samples are heated for 3 min at 70 degrees C just prior to loading. If this is not done, the oligos will not be fully denatured and this may result in a smeared background.

(3) Be sure to vigorously flush urea out of sample wells just prior to loading the sample. Urea will continually seep from the gel into the well. Urea is very dense and will force the sample into a ball or out of the well.

(4) If the sample volume is over 10 microliters, this may result in smearing.

(5) If ultrapure water (18 megaOhm) is not used, smeary bands and high background may result.

(6) Check to make sure the running and sample buffers are prepared and diluted correctly.

For the purpose of recovering and solubilizing nucleic acids in alcohol, can TAE running buffer be used instead of TBE running buffer when running a pre-cast TBE gel?

A TAE running buffer used in conjunction with a TBE gel would create high current and may lead to gel hydrolysis. Also, the acetate ion in TAE migrates faster than the borate ion in TBE buffer, and can create fuzzy bands. It is possible to try soaking the TBE gel (run in TBE buffer) in the TAE buffer following the run and then eluting from that. Since nucleic acids diffuse out of the gel relatively slowly, this may work.

What are the smallest fragment sizes that can be visualized on TBE gels?

On the Invitrogen 10% TBE gel, the 51-bp marker from the Boehringer Mannheim DNA MW V Kit can be clearly visualized. On the Invitrogen 20% TBE gel, the 18- and 12-bp markers from the same kit can be clearly seen at 2 micrograms total DNA loaded.

What is the recommended maximum sample loading volume for Novex TBE and TBE-Urea gels?

The recommended maximum sample loading volumes for Novex TBE and TBE-Urea gels are shown below:

10-well gels: 25 µL
12-well gels: 20 µL
15-well gels: 15 µL