missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE Gels, 8%

Novex™ TBE Gels 8% are designed to run on the XCell SureLock™ Mini-Cell and provide high-resolution analysis of restriction digests and PCR products.

Supplier:  Invitrogen™ EC62152BOX

Encompass

 View more versions of this product

Catalog No. EC62152BOX


Only null left

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.

Description

Description

Novex™ TBE Gels 8% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 25–220 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium­ bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results

Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

Specifications

Chemical Identifiers

• Catalog number refers to a single box of 10 gels

Store at 4°C.
TBE
10 Gels/Box
Wet Ice
XCell SureLock Mini-Cell
8%
25 to 220 bp
Double-stranded DNA (dsDNA)
TBE Gel

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
8%
25 to 220 bp
10 Gels/Box
Wet Ice
XCell SureLock Mini-Cell
Novex TBE Gels, 8%, 12-well
TBE
12-well
Double-stranded DNA (dsDNA)
TBE Gel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions