Learn More
Invitrogen™ Human alpha Synuclein ELISA Kit
ELISA
Supplier: Invitrogen™ KHB0061
Description
The Human alpha Synuclein ELISA quantitates Hu alpha Synuclein in human cell and tissue lysates. The assay will exclusively recognize both natural and recombinant Hu alpha Synuclein. Principle of the method The Human alpha Synuclein solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. These proteins are abundant in the brain and play a role in regulating synaptic transmission and membrane trafficking. Alpha- and beta-synuclein have been shown to selectively inhibit phospholipase D2, suggesting that they may also be involved in signaling pathways. Defects in the SNCA gene, which encodes alpha-synuclein, have been implicated in the pathogenesis of Parkinson's disease. Additionally, alpha-synuclein peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Multiple alternatively spliced transcripts encoding different isoforms have been identified for this gene. Diseases associated with SNCA include Parkinson Disease 1, Autosomal Dominant and Dementia, Lewy Body.Chemical Identifiers
| 0.23 to 15 ng/mL | |
| 6.4% | |
| Colorimetric Microplate Reader |
| 0.2 ng/mL | |
| 5.7 | |
| 6622 |
Specifications
| P37840 | |
| 0.2 ng/mL | |
| HRP | |
| Cell Lysates | |
| Colorimetric Microplate Reader | |
| NACP,PARK1,PARK4,PD1 | |
| 6.4% | |
| Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| α-Synuclein | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.23 to 15 ng/mL | |
| 1/56 to 100 ng/mL, 1.56 to 100 ng/mL | |
| ELISA | |
| Human | |
| 6622 | |
| 4 hr. | |
| 5.7 | |
| HRP | |
| 96 Tests | |
| Cell Lysate 10 μL | |
| alpha Synuclein | |
| 4 hrs |