missing translation for 'onlineSavingsMsg'
Learn More

Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Rabbit)

Catalog No. WP20007
Encompass
Change view
Click to view available options
Quantity:
100 mL (2 Bottles)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WP20007 100 mL (2 Bottles)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WP20007 Supplier Thermo Fisher Scientific Supplier No. WP20007
Only null left

Pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in a proprietary stabilizing reagent.

A goat anti-rabbit polyclonal alk-phos conjugate, this Secondary antibody is compatable with both chemiluminescent and chromogenic methods of immunodetection, and will identify antigens bound with a primary rabbit antibody.

Order Info

Shipping Condition: Wet Ice

Specifications

Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Quantity 100 mL (2 Bottles)
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store in refrigerator (2–8°C).
Form Liquid
Isotype IgG
Reactivity Rabbit
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.