missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PVDF/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm

Catalog No. LC2002
Encompass
Change view
Click to view available options
Quantity:
20 membrane/filter paper sandwiches
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2002 20 membrane/filter paper sandwiches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2002 Supplier Invitrogen™ Supplier No. LC2002
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pre-cut and pre-assembled—ready for protein blotting/transfer

Twenty membrane/filter paper sandwiches are provided, for 20 transfers. A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces

Applications Western transfer, protein sequencing, amino acid analysis, solid phase assay systems
Dimensions 8.3 x 7.3cm
Pore Size 0.2μm
Binding capacity 50 to 150 μg/cm2 for large, globular proteins; >150μg/cm2 for smaller peptides.

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Length (Metric) 8.3 cm
Width (Metric) 7.3 cm
Quantity 20 membrane/filter paper sandwiches
Format Sandwich
Material PVDF
Pore Size 0.2 μm
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Dimensions (LxW) 7.3 cm x 8.3 cm
Thickness 0.8 mm
Product Line Novex
Show More Show Less
How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Why must Invitrolon PVDF membranes be dried before staining?

Drying the PVDF membrane reduces the background staining that can occur with wet membranes. A dry PVDF membrane is very hydrophobic and doesn't wet well, but the areas where proteins are bound are more easily saturated and stainable.

This will not work with nitrocellulose membranes, and will only work with PVDF membranes stained for a brief period; staining beyond the recommended time will only increase the background and reduce the detectability.

How do I destain proteins on a PVDF membrane that were stained with SimplyBlue SafeStain?

After staining with SimplyBlue SafeStain, use deionized water for the less strongly retained protein bands on the PVDF membrane.

Increasing methanol or ethanol concentrations up to 70% should destain any remaining bands. You can leave the membrane in the destain indefinitely.

Can I use ethanol instead of methanol in the NuPAGE transfer buffer (and in transfer buffers in general)?

We find that including ethanol in the transfer buffer is just as effective as including methanol. You may use either of these alcohols at an equivalent concentration.

How can I reduce high background on PVDF Western blots?

PVDF membranes require more stringent blocking steps. This can be achieved by increasing the concentration of the blocking reagent 2 to 5 fold, increasing the blocking time, and performing the procedure at 37 degrees C. Blocking agents bind to unoccupied sites to prevent background staining and also to membrane-bound proteins, thus reducing non-specific interactions with the primary antibody. We offer WesternBreeze Immunodetection kits with blocking reagents and Pierce Fast Western Blot kits that have been pre-optimized to give low-background blots. Other examples of blocking agents are nonfat dry milk, BSA, and casein, Starting Block, and SuperBlock.

What are some causes of poor Western transfer efficiency with a PVDF membrane?

There are three main reasons why this may occur.

1) The membrane may not be properly treated prior to transfer. Make sure that the membrane is pre-wetted with a polar organic solvent such as methanol or ethanol. Failure to do so will prevent proteins from forming hydrophobic interactions with the membrane.

2) There may be poor gel-to-membrane contact. Ensure that the filter paper and blotting pads are well saturated with transfer buffer, taking care to remove any bubbles during the assembly of the membrane sandwich. The gel/membrane sandwich must fit securely in the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with new ones.

3) Overcompression of the gel: a good indication of overcompression is if the gel has been excessively flattened. In the event that the sandwich is overcompressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and membrane.

After transferring onto a PVDF membrane, what is the best way to store the membrane for future probing?

We recommend air drying the PVDF membrane and placing it in an envelope, preferably on top of a supported surface to keep the membrane flat. It can be stored indefinitely at ≤80 degrees C. Right before probing, we recommend re-wetting the membrane with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with the blocking step.

Which transfer buffers are recommended for peptide (N-terminal) sequencing?

Use non-glycine based buffers such as the NuPAGE Invitrogen Transfer buffer, CAPS, or 1/2X TBE transfer buffer.

What parameters can affect Western blotting transfer results?

SDS:
-Mobility out of the gel: increases mobility as SDS imparts negative charge to a protein and maintains it in a soluble state.
-Binding to membrane: reduces binding to nitrocellulose due to decreased hydrophobicity of the protein.
-Detection: may affect antigenicity of some proteins.

Alcohol in transfer buffer (i.e., methanol up to 20%):
-Mobility out of the gel: decreased; reduces pore size of gel.
-Binding to membrane: improves binding to nitrocellulose; removes SDS from proteins resulting in improved hydrophobic interactions.

Field strength (V/cm):
-Mobility out of the gel: field strength is the driving force of elution; if too low, sample is not completely transferred out of gel.
-Binding to membrane: if too high, sample passes through membrane without binding.

Transfer buffer:
-Mobility out of the gel: decreased if high conductivity and low resistance limit V/cm due to excessive heat generation; decreased if buffer pH is close to pI of native protein.

Membrane:
-Detection: PVDF and nylon require more stringent blocking conditions than nitrocellulose.

Gel:
-Mobility out of the gel: increases with thinner gels or larger gel pore size.

How can I reduce background bands in my Western blot?

Optimize the concentration of primary and secondary antibodies. In some cases, increasing the concentration of blocking agent (BSA or non-fat dry milk) or usiing an alternative blocking solution such as Starting Block or SuperBlock may reduce background signal. After incubation with the primary antibody, wash at least 2 times with TBST (include 0.5 M NaCl in one or more of the wash steps). Avoid Nonidet P40 or Triton X-100 in buffers because protein detection is decreased when these detergents are used.

How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

I stained my PVDF membrane with Coomassie Blue and am seeing strange swirly patterns in the background. What is the cause for this?

The most probable cause of the swirled artifact is the way the membrane is moved around in the staining solution. Using an orbital shaker can cause this effect. To eliminate these background patterns, we recommend switching to a rocker type shaker or combination of rocker and reciprocal motions to ensure even sloshing of the staining solution over the membrane.

I performed a western transfer onto a PVDF membrane and the transfer efficiency was very poor. Can you offer some tips?

Here are possible causes and solutions:

- The membrane may not be properly treated prior to transfer: Make sure that the membrane is pre-wetted with a polar organic solvent such as methanol or ethanol.
- There may be poor gel to membrane contact: Ensure that the filter paper and blotting pads are well saturated with transfer buffer, taking care to remove any bubbles during the assembly of the membrane sandwich. The gel/membrane sandwich must fit securely in the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones.
- Over-compression of the gel: A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and the membrane.

Are your PVDF and nitrocellulose membranes compatible with the Li-COR instrument?

Yes, both our PVDF and nitrocellulose membranes are compatible with the Li-COR instrument.

I am planning on doing a dot blot and my sample contains acetonitrile. Can your PVDF or nitrocellulose membrane withstand acetonitrile?

Our PVDF can tolerate acetonitrile but our nitrocellulose cannot.

How can I reduce background on my PVDF western blots?

PVDF membranes require more stringent blocking steps. This can be achieved by increasing the concentration of the blocking agent 2-5 fold, increasing the blocking time, and performing the procedure at 37 degrees C. Blocking agents bind to unoccupied sites to prevent background staining and also to membrane-bound proteins, reducing non-specific interactions with the primary antibody. Examples of blocking agents are nonfat dry milk, BSA, and Casein.


For Research Use Only. Not for use in diagnostic procedures.