missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ProBond™ Purification System
Designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence
Supplier: Invitrogen™ K85001
Includes: 6 x 2mL resin columns and buffers for native and denaturing purification; 12mL ProBond pre-charged resin
Description
- Utilizes proprietary ProBond nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions
His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
| Suspension | |
| Six 2-ml resin columns and buffers for native and denaturing purification. Twelve milliliters of ProBond™ pre-charged resin. Store at +4°C. All reagents are guaranteed stable for 6 months when properly stored. | |
| 6 purifications |
| ProBond™ Purification System | |
| His Tag | |
| ProBond |
For Research Use Only. Not for use in diagnostic procedures.