missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pNPP Powder
Description
May be used as a substrate for alkaline phosphatase (AP) in enzyme immunoassays.
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping condition: Wet ice
Specifications
Specifications
| Content And Storage | Store at 2-8°C |
| Detection Method | Colorimetric |
| Product Type | pNPP Powder |
| Substrate | pNPP |
| Substrate Properties | Chemical Substrate |
| Substrate Type | AP (Alkaline Phosphatase) Substrate |
| Quantity | 6 x 1 mL |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.