missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen⢠NuPAGE⢠Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007
Provide separation of large molecular weight proteins when used with NuPAGE Tris-Acetate SDS Running Buffer
Supplier: Invitrogen⢠LA0050
Includes: 20X NuPAGE Tris-Acetate SDS Running Buffer (500mL), 10X NuPAGE Sample Reducing Agent (250μl), NuPAGE Antioxidant (15mL), 4X NuPAGE LDS Sample Buffer (10mL)
Description
- NuPAGE Novex Tris-Acetate gels can also be run with Tris-Glycine Native Running Buffer to resolve native proteins more effectively than Tris-Glycine gel system
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
- Recycled Content
Specifications
NuPAGE Tris-Acetate Gel | |
SDS PAGE Gel | |
Buffer Kit |
NuPAGE⢠Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels) contains: ⢠20X NuPAGE⢠Tris-Acetate SDS Running Buffer (500 ml): Store at 4°C to 25°C ⢠10X NuPAGE⢠Sample Reducing Agent (250 μl): Store at 2°C to 8°C ⢠NuPAGE⢠Antioxidant (15 ml): Store at 4°C ⢠4X NuPAGE⢠LDS Sample Buffer (10 ml): Store at 4°C to 25°C |
|
NuPAGE⢠| |
1 kit |
Quick Reference: NuPAGE Tris-Acetate Mini Gels
Version: D
File size: 1 MB
For Research Use Only. Not for use in diagnostic procedures.