missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cell Lysis Buffer II DFS Item

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications.

Supplier:  Invitrogen™ FNN0021

Catalog No. FNN0021

Only null left

Description

Description

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications. The buffer is used for total protein extraction and utilizes detergent-based lysis, eliminating the need for mechanical cell disruption. The formulation helps retain the protein structure and function needed for enzyme assays or immunoassays.

Buffer formulation: 50 mM Tris, pH 7.4, 250 mM NaCl, 5 mM EDTA, 50 mM NaF, 1 mM Na3VO4, 1% detergent, 0.02% NaN3

This cell lysis buffer does not contain protease inhibitors and should be supplemented with 1 mM PMSF and protease inhibitor cocktail (Cat. No. A32955) just prior to use.

Order Info

Shipping condition: Dry ice

Specifications

Specifications

Store in freezer (-5 to -30°C).
Cell Lysis Buffer
100 mL
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.