missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm DFS Item

Ideal for blotting proteins and nucleic acids

Supplier:  Invitrogen™ LC2001

Catalog No. LC2001

Only null left
Explore available promotions

Description

Description

The 0.45μm pore size is ideal for the transfer of low molecular weight proteins (<20kDa) and nucleic acids (<300bp).

 

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Contain no support fabrics or detergents
  • Are compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, fluorescence, and radiolabeling
  • Provide high-sensitivity with low background
  • Are supplied in a pre-cut, pre-assembled membrane/filter paper sandwich for convenience
  • A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces

 

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

8.3 cm
Nitrocellulose Pre-Cut Blotting Membrane, 0.45μm pore size
Sandwich
0.45 μm
Room Temperature
0.8 mm
7.3 cm
20 membrane/filter paper sandwiches
Nitrocellulose
Store at room temperature.
7.3 cm x 8.3 cm
Novex
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.