missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP3 Human Matched Antibody Pair

Catalog No. CHC1543
Encompass
Change view
Click to view available options
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CHC1543 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CHC1543 Supplier Invitrogen™ Supplier No. CHC1543

Includes

Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.

Requires

Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).

Matched Antibody Pair

The Human MMP3 Antibody Pair Kit comes with pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human MMP3. Buffer reagents needed to complete the ELISA reaction are sold separate as a Buffer Kit for Antibody Pairs (Catalog No. CNB0011). The Assay Buffer included in this Buffer Kit can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.

Order Info

Shipping Condition: Wet Ice

Specifications

Conjugate HRP
Gene Accession No. P08254
Gene Alias CHDS6, MGC126102, MGC126103, MGC126104, MMP-3, SL-1, STMY, STMY1, STR1
Gene Symbols MMP3
Quantity 10 x 96 Tests
Regulatory Status RUO
Gene ID (Entrez) 4314
Includes Capture (coating) Antibody, Detection Antibody, Standard, Streptavidin-HRP
Content And Storage 2°C to 8°C
Product Type ELISA Kit
For Use With (Equipment) Colorimetric Microplate Reader
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.