Learn More
Invitrogen™ Human MMP2 ELISA Kit
ELISA
$804.00 - $1412.00
Specifications
| Accession Number | P08253 |
|---|---|
| Assay Range | 0.78 to 50 ng/mL |
| Assay Sensitivity | <1 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Description
The Human Matrix MetalloProteinase 2 (Hu MMP2) ELISA quantitates Hu MMP2 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MMP2. Principle of the method The Human MMP2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.Order Info
Shipping Condition: Dry Ice
Specifications
| P08253 | |
| <1 ng/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CLG4,CLG4A,MMP-2,MMP-II,MONA,TBE-1 | |
| 5.6% | |
| Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| MMP-2 | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MMP2 | |
| 4 hrs |
| 0.78 to 50 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| Colorimetric Detection | |
| 4313 | |
| 4 hr. | |
| 3.3 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
For Research Use Only. Not for use in diagnostic procedures.