Learn More
Invitrogen™ IL-6 Rat Matched Antibody Pair
Description
Includes
Antibody Pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Requires
Buffer reagents required to complete reaction available separately (Cat. No. CNB0011).
The Rat IL-6 Antibody Pair Kit comes with pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat IL-6. Buffer reagents needed to complete the ELISA reaction are sold separate as a Buffer Kit for Antibody Pairs (Catalog No. CNB0011). The Assay Buffer included in this Buffer Kit can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Specifications
Specifications
| Conjugate | HRP |
| Gene Accession No. | P20607 |
| Gene Alias | ILg6, Ifnb2 |
| Gene Symbols | Il6 |
| Quantity | 10 x 96 Tests |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 24498 |
| Includes | Capture (coating) Antibody, Detection Antibody, Standard, Streptavidin-HRP |
| Target Species | Rat |
| Content And Storage | 2°C to 8°C |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.