Learn More
Invitrogen™ Human IL-8 ELISA Kit, Ultrasensitive
Description
Includes
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
The Human Interleukin 8 (IL-8) ELISA quantitates Hu IL-8 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-8. Principle of the method The Human IL-8solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| Accession Number | P10145 |
| Assay Range | 0.39 to 25 pg/mL |
| Assay Sensitivity | <100 fg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | Colorimetric |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3576 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.