missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IFN gamma ELISA Kit

Catalog No. KMC4022
Encompass
Change view
Click to view available options
Quantity:
2 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KMC4022 2 x 96 Tests
KMC4021 96 Tests
KMC4021C 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. KMC4022 Supplier Invitrogen™ Supplier No. KMC4022

Includes

Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

ELISA

The Mouse Interferon Gamma (Ms IFNγ) ELISA quantitates Ms IFNγ in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IFNγ. Principle of the method The Mouse IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.

Order Info

Shipping Condition: Room Temperature

Specifications

Accession Number P01580
Assay Range 47 to 300 pg/mL
Assay Sensitivity <2 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 15978
Gene Symbol IFN-g,Ifg
Incubation Time 4 hr.
Interassay CV 6.5%
Intraassay CV 3.2
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene IFN-α
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 25 μL, Serum 25 μL, Supernatant 25 μL
Storage Requirements 2°C to 8°C
Target IFN gamma
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.