Learn More
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, regular size
GREENER_CHOICE
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins.
Supplier:  Invitrogen™ IB23001
Encompass
 View more versions of this product
Catalog No. IB23001
Only null left
Description
Description
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This regular-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels or E-PAGE 48 or 96 gels.
Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.
Specifications
Chemical Identifiers
Transfer Stack
0.2 μm
13.5 x 8 cm
Nitrocellulose
Store at room temperature.
iBlot
Specifications
13.5 cm
10 Stacks
Nitrocellulose
Store at room temperature.
iBlot
8 cm
Transfer Stack
0.2 μm
13.5 x 8 cm
10 Blots
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.