missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HRP-Conjugate Diluent/Stabilizer

Catalog No. 002018
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
002018 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 002018 Supplier Invitrogen™ Supplier No. 002018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for use as conjugate diluent in enzyme immunoassays

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

Specifications

Content And Storage Store at 2-8°C
Product Type Conjugate
Quantity 1 L
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.