missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel Loading Tips, standard, round

Catalog No. LC1001
Encompass
Change view
Click to view available options
Quantity:
200/ pkg.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC1001 200/ pkg.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC1001 Supplier Invitrogen™ Supplier No. LC1001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For easy and accurate loading of samples

Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 μL.

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Specifications

Content And Storage Contains 1 pack (200/pack) of Standard, Round gel loading tips.
Description Standard round Tips
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment) Universal Fit
Type Gel Loading Tip
Filtered No
Quantity 200/ pkg.
Shipping Condition Room Temperature
Sterility Non-sterile
Volume (Metric) 200 μL
Do you offer gel loading tips to load protein gels?

We offer Invitrogen Gel Loading Tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable. Three varieties are offered: standard round tips, flat gel tips, and Eppendorf round gel tips.

For Research Use Only. Not for use in diagnostic procedures.