missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel-Dry™ Drying Solution

Modulates the rate of water release to ensure crack-free gels

$110.00 - $470.00

Specifications

Buffer Drying Solutions
Content And Storage Store at room temperature.
For Use With (Equipment) DryEase™ Mini-Gel Drying System
Gel Size Mini
Product Line Novex
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Buffer Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Buffer Price Quantity & Availability  
LC4025
Encompass
View Documents Promotion Details
Invitrogen™
LC4025
500 mL
Each for $110.00
Only null left
 
LC40254
Encompass
View Documents Promotion Details
Invitrogen™
LC40254
8 x 500 mL
Each for $470.00
Only null left
 
Description

Description

  • Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System which is a simple, easy-to-use kit for drying gels evenly using passive evaporation
  • The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air
  • Consequently, drying is even and cracking is minimized
  • The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels
  • With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking

Applications

  • Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

Drying Solutions
DryEase™ Mini-Gel Drying System
Novex
Room Temperature
Store at room temperature.
Mini
Drying Solution
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.