missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel-Dry™ Drying Solution

Catalog No. LC40254
Encompass
Change view
Click to view available options
Volume (Metric) Buffer:
500 mL
8 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Buffer
LC40254 8 x 500 mL
LC4025 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC40254 Supplier Invitrogen™ Supplier No. LC40254

Modulates the rate of water release to ensure crack-free gels

  • Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System which is a simple, easy-to-use kit for drying gels evenly using passive evaporation
  • The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air
  • Consequently, drying is even and cracking is minimized
  • The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels
  • With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking

Applications

  • Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage

Order Info

Shipping condition: Room Temperature

Specifications

Buffer Drying Solutions
Content And Storage Store at room temperature.
For Use With (Equipment) DryEase™ Mini-Gel Drying System
Gel Size Mini
Product Line Novex
Type Drying Solution
Quantity 8 x 500 mL
Shipping Condition Room Temperature
Volume (Metric) Buffer 8 x 500 mL
What is the composition of the Gel-Dry Drying solution?

The Gel-Dry Drying solution has a proprietary composition.

Is the Gel-Dry Drying solution compatible with the enhancers used in autoradiography?

Yes, it is compatible. You need to put the gel in the enhancer solution, wash it and then proceed with the gel drying solution. For detection of signal using 32P, the cellophane is fine. For 35S, the upper layer of cellophane may need to be removed (which is possible) depending on the enhancer being used. For detection of 14C, there is no need to remove the cellophane as it will pass through.

For Research Use Only. Not for use in diagnostic procedures.