missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGF2 ELISA Kit

ELISA

Supplier:  Invitrogen™ KHG0021

Encompass

 View more versions of this product

Catalog No. KHG0021


Only null left

Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Human FGF-b ELISA Kit quantifies human Fibroblast Growth Factor basic (FGF-b) in serum, plasma, buffered solution, or cell culture medium. The assay will recognize both the natural 18 kDa isoform and recombinant FGF basic-146 and FGF basic-157. Principle of the method The Human FGF-b solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Identifiers

P09038
7 pg/mL
ELISA
Human
Colorimetric Microplate Reader
BFGF,FGF-2,FGFB,HBGF-2
5.5%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
FGF-Basic
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
FGF2
4 hrs
15.6 to 1000 pg/mL
HRP
Plasma, Serum, Supernatant
Colorimetric Detection
2247
4 hr.
4.3
HRP
RUO
2°C to 8°C
1 hrs 20 mins

Specifications

P09038
7 pg/mL
ELISA
Human
Colorimetric Microplate Reader
BFGF,FGF-2,FGFB,HBGF-2
5.5%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
FGF-Basic
RUO
2°C to 8°C
1 hrs 20 mins
15.6 to 1000 pg/mL
HRP
Plasma, Serum, Supernatant
Colorimetric Detection
2247
4 hr.
4.3
HRP
96 Tests
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
FGF2
4 hrs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.