missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Empty Gel Cassettes, mini, 1.0 mm

Catalog No. NC2010
Encompass
Change view
Click to view available options
Quantity:
25⁄pkg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC2010 25⁄pkg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC2010 Supplier Invitrogen™ Supplier No. NC2010

For pouring gels

All cassettes are sealed on three sides to prevent leaking. Available in 1.0mm thickness and for use with seven comb types (1-well, 2-well, 5-well, 10-well, 12-well, 15-well, and 2D-well). Pre-sealed with pre-taped slot so bulky casting stand is not necessary.

Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Content And Storage Contains a pack (25/pack) of empty, disposable, plastic gel cassettes (1.0 mm). Store at room temperature.
For Use With (Equipment) Universal
Gel Size Mini (8 x 8 cm)
Product Line Novex
Type Empty Gel Cassette
Quantity 25⁄pkg
Shipping Condition Room Temperature
Are the Invitrogen empty gel cassettes and combs RNase-free?

No. The cassettes and combs are not RNase free. There is no sterility QC test for RNase activity. It may be possible to treat with RNaseZAP, or another RNase decontamination solutions followed by a thorough rinse with RNase-free water.

Do you offer empty cassettes and combs for casting Mini and Midi protein gels?

We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).

- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).

For Research Use Only. Not for use in diagnostic procedures.