Learn More
Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini
Catalog No. NC3002
Encompass
Change view
Click to view available options
Thickness (Metric):
1.0 mm
Catalog No. Thickness (Metric)
NC3002 1.0 mm
1 options
Catalog No. NC3002 Supplier Invitrogen™ Supplier No. NC3002
Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)
These combs are for use when pouring special gel types and are available in two thicknesses (1.0mm and 1.5mm) and seven well formats (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
For Use With (Equipment) Mini-size Empty Gel Cassettes
Type 2D-well Empty Gel Cassette Comb
Thickness (Metric) 1.0 mm
For Research Use Only. Not for use in diagnostic procedures.