missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EKMax™ Enterokinase

Catalog No. E18001
Encompass
Change view
Click to view available options
Quantity:
1000 Units
250 Units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E18001 250 Units
E18002 1000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E18001 Supplier Invitrogen™ Supplier No. E18001

Includes

EKMax Enterokinase is provided with 10X buffer.

Recombinant preparation of the catalytic subunit of bovine enterokinase

  • Recognizes the sequence Asp-Asp-Asp-Asp-Lys and cleaves peptide bond after lysine residue
  • Enzyme can be used to cleave any fusion protein that carries this peptide sequence
  • Well-suited to removal of fusion tags from recombinant proteins
  • Unit Definition: One unit of EKMaxis amount of enzyme required to digest 20μg of thioredoxin-CAT fusion protein to 90% completion in 16 hours at 37°C; One EKMaxunit is equivalent to ∽190 trypsinogen activation units

GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Specifications

Product Type Enterokinase
Content And Storage 250 or 1000 units of EKMax™ is provided with 10X buffer. Store at -20°C. Guaranteed stable for 6 months when stored properly.
Species Cattle/Bovine
Quantity 250 Units
Product Line EKMax
What is the molecular weight of the EKMax enterokinase enzyme?

EKMax enterokinase is a clone of the catalytic subunit of enterokinase expressed in the yeast Pichia pastoris. The calculated molecular weight of the protein is 26.3 kDa, but it contains three sites for asparagine-linked glycosylation. The apparent molecular weight of 43 kDa is consistent with previous observations (LaVallie et al., 1993) and is assumed to be because of N-linked glycosylation.
Reference: LaVallie, E.R., Rehemtulla, A., Racie, L.A.,Diblasio, E.A., Ferenz, C., Grant, K.L. Light, A., and McCoy, J.M. (1993). Cloning and functional expression of a cDNA encoding the catalytic subunit of bovine enterokinase. J.Biol.Chem. 268, 23311-23317.

Will DTT, Triton X-100 detergent, Tween 20 detergent, Thesit, calcium chloride, sodium chloride, or SDS affect the efficiency of Enterokinase (EKMax) enzyme cleavage?

Enterokinase is active in buffers containing up to 1 mM DTT, 0.1% Triton X-100 detergent, 0.1% Tween 20 detergent, and 0.1% Thesit. It is recommended to have 10mM Tris pH 8.0 and 10 mM calcium chloride in the buffer. Enterokinase is inhibited by sodium chloride and SDS.

Should Enterokinase be resuspended in a buffer containing 50% glycerol to protect the protein from freeze/thaw cycles?

Freeze thaw has a minimal effect on the activity of Enterokinase. The addition of glycerol is not necessary but can make handling of the enzyme easier.

How specific is cleavage by EKMax Enterokinase? Are there any alternate cleavage sites for the enzyme?

Enterokinase cleaves after the sequence (Asp)4-Lys.

It has been proposed that the active center of enterokinase possesses a distinctive cationic subsite that binds -(Asp)4. Enterokinase is highly specific and tolerates very few changes to its recognition site. If the ionic charge of the recognition site is preserved, enterokinase will recognize the site, but the rate of hydrolysis of the peptide bond will be reduced (Light and Janska, 1989). The four aspartyl residues act as a signal for enterokinase cleavage. It has been reported that with only three aspartyl residues the rate of hydrolysis is reduced. Two aspartyl residues preceding the lysyl residue are the minimum number of acidic residues needed to maintain specificity (Maroux et al., 1971). Non-specific cleavage by enterokinase may occur in the cases described above, but this is usually alleviated by reducing the amount of enzyme used.

What products do you offer for enzymatic cleavage of fusion tags from recombinant proteins?

We offer the following products:

-AcTEV Protease (Cat. Nos. 12575015, 12575023)
-EKMax Enterokinase (Cat. Nos. E18001, E18002)
-SUMO Protease (Cat. No. 12588018)

For Research Use Only. Not for use in diagnostic procedures.