missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Buffer Core Repair Kit for XCell SureLock™ Mini-Cell

Catalog No. EI9021
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EI9021 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EI9021 Supplier Invitrogen™ Supplier No. EI9021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Includes a platinum wire, wire insulation tubing, banana plug, hex nut, and washer.

For the XCell SureLock™ Mini-Cell Blot Module

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment) XCell SureLock™ Mini
Gel Size Mini
Product Line XCell SureLock
Type Mini-Cell Buffer Core Repair Kit
Quantity 1 kit
Shipping Condition Room Temperature
Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

For Research Use Only. Not for use in diagnostic procedures.