Learn More
Invitrogen™ Human beta Amyloid (1-40) PTD Recombinant Protein
Description
Human beta Amyloid (1-40) PTD Recombinant Protein for Bioactivity
Specifications
Specifications
| Accession Number | P05067 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 351 |
| Molecular Weight (g/mol) | 4.3 kDa |
| Name | Human beta Amyloid (1-40) PTD |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Frequently Asked Questions (FAQs)
Recommendations for Beta Amyloid Peptide Reconstitution:
(1) To induce peptide aggregation for neurotoxicity studies: The appearance of toxicity has recently been shown to correlate to the extent of beta sheet structure (S. Wang et al. [2001] J. Biol. Chem. 276(45):42027-42034). Recommended preincubation is: Dissolve the lyophilized peptide in 0.1% (v/v) trifluoroacetic acid in water at 10 mg/mL. Dilute the peptide to 0.5-1.0 mg/mL with PBS (without Ca2+). Incubate at 25°C for 24-48 hrs (24-36 hrs is usually sufficient). Neurotoxic activity is usually observed at 30-100 micrograms/mL.
(2) To prepare peptide for studies which require minimal peptide aggregation: Dissolve the peptide at a concentration of 1 mg/mL in 100% HFIP (1,1,1,3,3,3-hexafluoro-2-propanol [Sigma-Aldrich Cat. No. 325244, 99.8% ACS reagent grade]). Incubate at RT for 2 hours. During the incubation, vortex the peptide solution several times at moderate speed, allowing the HFIP to cover as much of the surface area as possible. Dry down the HFIP/peptide solution under a gentle stream of nitrogen gas. Continue drying for an additional 10 minutes. Cap vial immediately. Resuspend the peptide in 100% DMSO. Incubate the peptide plus DMSO for 12 minutes at RT with periodic vortexing at moderate speed. Add 10 microliters of this DMSO/peptide solution dropwise to 10 mL of BSAT-DPBS (see formulation below) while vortexing at moderate speed. Peptides prepared in this manner have been used as standards in ELISA for the detection of beta amyloid in biological samples. When using this peptide in ELISA, it is important that the assay buffer used with the peptide standards has the same composition as the samples under investigation.
Buffer Formulations: DPBS Solution (10X Stock) (Gibco#14200-075) Dulbecco's PBS (DPBS w/o Mg2+, Ca2+) BSAT-DPBS Solution: 1X DPBS, pH 7.4 5% BSA 0.03% Tween-20
Note: The inclusion of a protease inhibitor (i.e. AEBSF [Sigma Cat. No. A-8456]) is recommended when the BSAT-BPBS solution is to be used for the analysis beta amyloid-containing biological samples. To prepare a 40 mM stock solution of AEBSF, add 100 mg of AEBSF to 10 mL DPBS, pH 7.4, containing 5% BSA. The stock solution should be diluted to a concentration of 1 mM AEBSF in BSAT-DPBS just prior to use.