missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABTS, solution

Catalog No. 002024
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
002024 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 002024 Supplier Invitrogen™ Supplier No. 002024

Suitable for use in immunoassays

May be used as a substrate for horseradish peroxidase (HRP) in ELISAs.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

Specifications

Content And Storage Store at 2-8°C
Detection Method Colorimetric
Product Type ABTS Solution
Substrate ABTS
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Quantity 500 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.