Learn More
MilliporeSigma™ Novagen™ Veggie BL21 (DE3) Singles™ Competent Cells
Catalog No. 712524
Change view
Click to view available options
Quantity:
22 Reactions
Catalog No. Quantity
712524 22 Reactions
1 options
Catalog No. 712524 Supplier MilliporeSigma™ Supplier No. 712524
Only null left
Protease-deficient hosts (BL21) for protein expression; Manufactured free of animal-derived media and components
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Specifications
Content And Storage ≤-70°C
Quantity 22 Reactions