missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Rosetta™ (DE3) pLysS Competent Cells

Catalog No. 709564
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709564 1 mL
709563 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 709564 Supplier MilliporeSigma™ Supplier No. 709564

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. T7 lysozyme expression suppresses basal T7 expression.

Rosetta™ host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta strains provide for “universal” translation which is otherwise limited by the codon usage of E. coli. The tRNA genes are driven by their native promoters. In Rosetta(DE3)pLysS, the rare tRNA genes are present on the same plasmids that carries the T7 lysozyme gene. DE3 indicates that the host is a lysogen of ™DE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.

Specifications

Content And Storage ≤-70°C
Quantity 1 mL