missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ RNase A Solution

Catalog No. 708563
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
708563 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 708563 Supplier MilliporeSigma™ Supplier No. 708563

Suitable for use in selective removal of RNA.

EMD Millipore Novagen™ RNase A Solution is suitable for RNA digestion in plasmid purification procedures.

  • Highly purified preparation of bovine pancreatic ribonuclease A
  • Pretreated to remove DNase I is suitable for RNA digestion in plasmid purification procedures
  • Supplied at concentration of 10mg/mL in 10mM Tris-HCl, 1mM EDTA, 50% glycerol, pH 7.5

Specifications

Concentration 10 mg/mL
Content And Storage Store at 15° to 30°C.
For Use With (Application) Selective removal of RNA
Packaging Type Plastic ampoule
Quantity 1 mL
Sample Source Bovine pancreatic ribonuclease A