missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ RiboJuice™ siRNA Transfection Reagent

Catalog No. 711154
Change view
Click to view available options
Quantity:
1 mL
300 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
711154 1 mL
711153 300 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 711154 Supplier MilliporeSigma™ Supplier No. MIL711154

missing translation for 'validMainframeMsg'

Delivers siRNA into mammalian cells for targeted gene suppression.

EMD Millipore Novagen™ RiboJuice™ siRNA Transfection Reagent is specifically optimized for siRNA delivery.

  • Formulated for minimal toxicity
  • Compatible for transfection of siRNA, plasmid DNA for normalizing transfection efficiency with reporter gene
  • Application:
    • Successfully used for gene suppression in variety of cell lines
    • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to -8°C
Quantity 1 mL