missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ pTK-neo DNA

Catalog No. 712843
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712843 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 712843 Supplier MilliporeSigma™ Supplier No. 712843

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for selection of stably transfected mammalian cell lines (G418). MilliporeSigma™; Novagen™ pTK-neo DNA has a minimal thymidine kinase (TK) promoter controls expression of the neomycin resistance gene.

  • TK promoter  facilitates selection for stable integration of both selection plasmid and cotransfected expression plasmid into transcriptionally active sites in the genome
  • Used as cotransfection plasmid with pTandem™-1 Mammalian Dual Expression Vector, pTriEx™ vectors or any mammalian expression vector

Specifications

Format Glass bottle
Product Type neo DNA
Content And Storage Store at -20°C. Ok to freeze.
Quantity 20 μg