missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Origami™ B (DE3) Competent Cells

Catalog No. 708374
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
708374 1 mL
708373 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 708374 Supplier MilliporeSigma™ Supplier No. 708374

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Origami B host strains carry the same mutations as the original Origami strain, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG.

Origami™ B host strains carry the same mutations in trxB and gor as the original Origami strains, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels by adjusting the concentration of IPTG. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner™, and Origami strains in one strain background. The mutations in trxB and gor are selectable on kanamycin and tetracycline, respectively; therefore, these strains cannot be used with plasmids that can only selected with kanamycin or tetracycline. These strains also include the lon and ompT deficiences of BL21, which increase protein stability. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL