missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ NovaCHOice™ Transfection Kit

Catalog No. p-7078313
Change view
Click to view available options
Quantity:
1 mL/Pk.
10 mL/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
726223 1 mL/Pk.
726224 10 mL/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 726223 Supplier MilliporeSigma™ Supplier No. 726223
Only null left

Developed for mammalian protein production in Chinese Hamster Ovary (CHO) derived cells.

  • Enhances expression in certain media formulations
  • Consists of animal-origin free components
  • Suitable for both transient and stable transfection
  • Packaging: Glass Bottle

Specifications

Content And Storage -20°C
Includes BacMagic-3 DNA, Insect GeneJuice Transfection Reagent, Sf9 Cells, BacVector™ Insect Cell Medium, Transfection Control Plasmid
Quantity 1 mL/Pk.
Form Liquid