missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ NanoJuice™ Core Transfection Reagent

Catalog No. 719003
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
719003 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 719003 Supplier MilliporeSigma™ Supplier No. 719003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to be used in concert with NanoJuice Transfection Booster.

  • Polycationic liposomal formulation
  • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to -8°C
Quantity 1 mL
Form Liquid