missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Lysonase™ Bioprocessing Reagent

Catalog No. 712304
Change view
Click to view available options
Quantity:
0.2 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712304 1 mL
712303 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 712304 Supplier MilliporeSigma™ Supplier No. 712304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy, optimized, ready-to-use blend of rLysozyme™ Solution and Benzonase Nuclease. MilliporeSigma™; Lysonase™ Bioprocessing Reagent contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme.

  • Benzonase Nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear)
  • Reduces extract viscosity and increases protein yield
  • Combined activities of rLysozyme and Benzonase Nuclease significantly increase protein extraction efficiency and facilitate downstream processing of protein extracts
  • For efficient protein extraction with BugBuster™ Protein Extraction Reagent, use 10μL Lysonase per 1g cell paste
  • For efficient protein extraction with PopCulture™ Reagent, add 2μL Lysonase per 1mL culture

Specifications

For Use With (Application) Protein Extraction, Protein Preparation
Quantity 1 mL
Product Type Bioprocessing Reagent
Content And Storage -20°C