missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ HMS174 (DE3) Competent Cells

Catalog No. 694534
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
694534 1 mL
694533 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 694534 Supplier MilliporeSigma™ Supplier No. 694534

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HMS174 strains provide high transformation efficiencies and the recA mutation in a K-12 background. Strain may stabilize certain target genes whose products may cause the loss of the DE3 prophage.

HMS174 strains provide the recA mutation in a K-12 background. Like BLR, these strains may stabilize certain target genes whose products may cause the loss of the DE3 prophage. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

Specifications

Efficiency Guaranteed efficiency: >5 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL